m-Cresol purple sodium salt

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

S0045-1G S0045-5G
TCIAS0045-1GEA 26.4 EUR
TCIAS0045-1G TCIAS0045-5G
m-Cresol purple sodium salt
m-Cresol purple sodium salt
Formule: C₂₁H₁₇NaO₅S
MDL Nummer: MFCD00010177
CAS nummer: 62625-31-4
EINECS: 263-656-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19ac168a98e4d1760efb8f5297c7fb43006d01cc&vid=19ac168a98e4d1760efb8f5297c7fb43006d01cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Red to Dark blue to Black powder to crystal
Lambda max. 434.0 to 437.0nm (KH2PO4・NaOH pH 7 buffer sol.)
Absorbance (E1%1cm) min. 350 (10mg/L, 434.0 to 437.0nm (KH2PO4・NaOH pH 7 buffer sol., 434.0 to 437.0nm)
Solubility in Water almost transparency
Drying loss max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR