Dispenser tips, ritips®
Leverancier: BIOZYM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
742100 742101 742102 742103 742104 742105 742108 742109 742110 742111 742112 742113 742114 742115 742116 742119
BZYM742100EA 0 EUR
BZYM742100 BZYM742101 BZYM742102 BZYM742103 BZYM742104 BZYM742105 BZYM742108 BZYM742109 BZYM742110 BZYM742111 BZYM742112 BZYM742113 BZYM742114 BZYM742115 BZYM742116 BZYM742119
Dispenser tips, ritips®
Pipettips
These dispenser tips are compatible with Ritter ripette®, Eppendorf Multipette® 4780 and many other dispensing systems.
Bioclean® tips are sterile and free from DNase, RNase and pyrogens.
Leveringsinformatie: The 25 ml and 50 ml tips are supplied with adapter.