Cryogenic vials, Cryovials®
Leverancier: BIOZYM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
710501 710502 710503 710504 710505 710506 710511 710512 710513 710514 710515 710516 710518 710521 710522 710523 710524 710525 710526
BZYM710501EA 0 EUR
BZYM710501 BZYM710502 BZYM710503 BZYM710504 BZYM710505 BZYM710506 BZYM710511 BZYM710512 BZYM710513 BZYM710514 BZYM710515 BZYM710516 BZYM710518 BZYM710521 BZYM710522 BZYM710523 BZYM710524 BZYM710525 BZYM710526
Cryogenic vials, Cryovials®
flesjes
These cryogenic vials made from PP are used for optimum storage of a variety of biological materials such as cell cultures, sera and blood components.
  • Gamma sterilised and DNA-, DNAse-, RNAse- and pyrogen free
  • Silicone sealing ring for safe and dense closure
  • Vertical lids for tight fingers
  • White, scratch-resistant label
  • Perfect sample visibility through clear vascular walls
  • Snap-in thread for all free-standing tubes