Cervical scrapers, Ayre spatula, plastic
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
440142 444142
231-2601EA 141 EUR
231-2601 231-2611
Cervical scrapers, Ayre spatula, plastic
Celschrapers
PP spatula with rounded edges. Sterile model is sterilised by ethylene oxide. Pack includes instructions for use.