IP Biopsy VI-cassettes, los
Leverancier: LEICA HISTOLOGY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
39LC-550-1 39LC-550-10 39LC-550-11 39LC-550-2 39LC-550-3 39LC-550-4 39LC-550-6 39LC-550-5 39LC-550-7 39LC-550-8 39LC-550-9
720-2279EA 108 EUR
720-2279 720-2471 720-2473 720-2475 720-2477 720-2479 720-2483 720-2481 720-2485 720-2487 720-2489
IP Biopsy VI-cassettes, los
Histologie-cassettes Cassettes voor biopsiebehandeling
ActivFlo Routine VI cassettes are processing cassettes featuring a patented lateral vent design for maximum fluid exchange. The increase in fluid exchange results in less incidence of unfixed or under-processed tissues and faster fixation/processing of tissue specimens.
  • One-piece cassettes
  • Frangible’ lids for easy removal
  • 45° writing surface
  • Offer <1% printing error rate
  • Available in several colours