Disposable weighing boats, anti-static, Sterilin®
Leverancier: STERLIN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WB30335 WB30338
SLINWB30335EA 0 EUR
SLINWB30335 SLINWB30338
Disposable weighing boats, anti-static, Sterilin®
Schuitjes Weegschuitjes
PS, diamond shaped.
  • Avoids the retention of powders by static electricity
  • Ideal for use with fine powders e.g. talcum powder
Raw material modified to significantly reduce the charge relaxation time in comparison to standard weigh boats. Majority of electrostatic charge dissipates within seconds when earthed on the balance pan.