Trousers for women, C.I., Design C, dark grey
Leverancier: FRISTADS KANSAS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
844941131C44 844941131C46 844941131C48 844941210C44 844941210C46 844941210C48 844941231C44 844941231C46 844941231C48 844941331C44 844941331C46 844941331C48 844941440C44 844941440C46 844941440C48 844941520C44 844941520C46 844941520C48 844941530C44 844941530C46 844941530C48 844941539C44 844941539C46 844941539C48 844941640C44 844941640C46 844941640C48 844941730C44 844941730C46 844941730C48 844941940C44 844941940C46 844941940C48 844941941C44 844941941C46 844941941C48 84494134 84494136 84494138 84494140 84494142 84494144 84494146 84494148 84494150
KWIN844941131C44EA 0 EUR
KWIN844941131C44 KWIN844941131C46 KWIN844941131C48 KWIN844941210C44 KWIN844941210C46 KWIN844941210C48 KWIN844941231C44 KWIN844941231C46 KWIN844941231C48 KWIN844941331C44 KWIN844941331C46 KWIN844941331C48 KWIN844941440C44 KWIN844941440C46 KWIN844941440C48 KWIN844941520C44 KWIN844941520C46 KWIN844941520C48 KWIN844941530C44 KWIN844941530C46 KWIN844941530C48 KWIN844941539C44 KWIN844941539C46 KWIN844941539C48 KWIN844941640C44 KWIN844941640C46 KWIN844941640C48 KWIN844941730C44 KWIN844941730C46 KWIN844941730C48 KWIN844941940C44 KWIN844941940C46 KWIN844941940C48 KWIN844941941C44 KWIN844941941C46 KWIN844941941C48 KWIN84494134 KWIN84494136 KWIN84494138 KWIN84494140 KWIN84494142 KWIN84494144 KWIN84494146 KWIN84494148 KWIN84494150
Trousers for women, C.I., Design C, dark grey
Werkkleding
Robust trousers for ladies, made from 65% polyester and 35% cotton, with soft nap on reverse side.
  • Fly with zip
  • Two front pockets, two back pockets with flap and velcro closure
  • Leg pockets with flaps and three tool pockets, leg pocket with mobile phone pocket and two pen pockets
  • Folding-ruler pocket, D-ring, inside knee pockets
Weight: 260 g/m²
Basic colour: Dark grey
Bestelinformatie: Trousers are available without any coloured accents or with 12 different accent colours.