Calcium chloride dihydrate, EMPROVE® EXPERT BP, Ph. Eur., JP, USP for biopharmaceutical production
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Calcium Chloride Dihydrate, Calcium dichloride
1.37101.5000 1.37101.9012 1.37101.9029
1.37101.5000EA 324 EUR
1.37101.5000 1.37101.9012 1.37101.9029
Calcium chloride dihydrate, EMPROVE® EXPERT BP, Ph. Eur., JP, USP for biopharmaceutical production
Calcium chloride dihydrate
Formule: CaCl₂·2H₂O
MW: 147.01 g/mol
Kookpunt: 1600
Smeltpunt: 176
Dichtheid: 1.85 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149613
CAS nummer: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660