Calcium chloride dihydrate, EMPROVE® ESSENTIAL E509, FCC, BP, Ph. Eur., JP, USP, SAFC®
Leverancier: MERCK PRODUCTION CHEMICALS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Calcium Chloride Dihydrate, Calcium dichloride
1.42002.5000 1.42002.9029 1.42002.9525 1.42002.1000
1.42002.5000EA 288 EUR
1.42002.5000 1.42002.9029 1.42002.9525 1.42002.1000
Calcium chloride dihydrate, EMPROVE® ESSENTIAL E509, FCC, BP, Ph. Eur., JP, USP, SAFC®
Calcium chloride dihydrate
pH value: 4.5 - 8.5 (50 g/l, H₂O, 20 °C)
Solubility: 1280 g/l
Formule: CaCl₂·2H₂O
Kookpunt: 1600 °C (1013 hPa)
Smeltpunt: 176 °C
Dichtheid: 1,85 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149613
CAS nummer: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660