VWR® Q Path®, Medium-acting decalcifiers
Leverancier: VWR Q-Path Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11028304.EA 172 EUR
11028304. 11028306.
VWR® Q Path®, Medium-acting decalcifiers
Kleurstoffen Microscopy and staining reagents
This medium-acting decalcifier contains a mixture of hydrochloric acids.
  • No formalin in the mixture
  • Decalcify larger tissues already fixated
  • Recommended volume is 20× the fragment volume
Ready to use for applications in histology and pathology laboratories.