VWR® Q Path®, Slow Acting Decalcifiers
Leverancier: VWR Q-Path Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11028303.EA 126 EUR
11028303. 11028305.
VWR® Q Path®, Slow Acting Decalcifiers
Kleurstoffen Microscopy and staining reagents
DC1 slow acting decalcifiers contain a mixture of formic acid and formalin.
  • Decalcify and fix small fragments (5 mm) in two to four hours
  • Recommended volume is 20× the fragment volume
Ready to use for applications in histology and pathology laboratories.