HPLC columns, HyPURITY™ Aquastar
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22505-254630 22505-154630 22505-052130 22503-104030 22503-154630 22503-013001 22503-012101 22503-153030
555-8155EA 1080 EUR
555-8155 555-8149 555-8085 555-8071 555-8077 555-8117 555-8116 555-8075
HPLC columns, HyPURITY™ Aquastar
Kolommen voor chromatografie
These columns are a silica-based polar end-capped and it is designed to provide superior retention of polar compounds and increased sensitivity in 100% aqueous conditions.
  • Ideal for structurally similiar analytes
  • Optimised for LC/MS applications