HPLC columns, HyPURITY™ C4
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
22405-254630 22405-154630
555-8114EA 1160 EUR
555-8114 555-8109
HPLC columns, HyPURITY™ C4
Kolommen voor chromatografie
These columns are ideal for the separation of structurally similar analytes.
  • Increased retention of polar compounds
  • Ideal for critical pairs analyses