Dekglaasjes, rond
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0111540 0111530 0111700 0111580 0111560 0111600 0111620 0111640 0111500 0111520 0111550 0111650
631-0692EA 85 EUR
631-0692 630-1597 631-0694 631-0669 631-0668 631-0670 631-0671 631-0672 631-0665 631-0666 631-0667 MARI0111650
Dekglaasjes, rond
Dekglaasjes
Round cover glasses made of pure white borosilicate D 263 ™ M glass, Hydrolytic Class I.
  • No bubbles or flaws
Suitable for in vitro diagnostic (IVD) applications.
Verpakking: 100 cover glasses per box with separate lid.
Order Now
Specificaties
Environmentally Preferable