Retention gap kits
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
290GU498 290GU499 260G498P
555-9200EA 78.3 EUR
555-9200 555-9199 555-9501
Retention gap kits
Slangen Chromatography Tubing
Use these gap kits to conveniently install a retention gap/guard column to your analytical column.
Leveringsinformatie: Each kit includes deactivated silica tubing, a miniunion, and five ferrules.