Gas cartridge filters, Super Clean™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60180-819 60180-820 60180-821 60180-822 60180-824 60180-825 60180-827 60180-826 60180-828 60180-829 60180-830
HYPE60180-819EA 268 EUR
HYPE60180-819 HYPE60180-820 HYPE60180-821 HYPE60180-822 HYPE60180-824 HYPE60180-825 HYPE60180-827 HYPE60180-826 HYPE60180-828 HYPE60180-829 HYPE60180-830
Gas cartridge filters, Super Clean™
Filters Gasfilters
These filters can be easily replaced without tools.
  • 99,9999% pure gas (or 6,0 grade) output
  • No carrier gas line contamination during cartridge change
  • Maximum pressure of 11 bar (160 psi)
  • Maximum flow of 7 L/min
Certificaten: TUEV approved.