Lege navulrekken
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2004-ER 2006-ER 2008-ER
732-1171EA 20.5 EUR
732-1171 732-1170 732-1169
Lege navulrekken
Pipette Tip Boxes and Racks
Save valuable space in the lab with the stackable Thermo Scientific™ ART™ pipette tip empty hinged racks.
  • Certified to be free of RNase, DNase, DNA, ATP, Endotoxin, Pyrogen and PCR Inhibitors
  • Front latch closes and locks, ensuring safe and secure storage throughout