PTFE/silicone discs, Tuf-Bond™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TS-12712 TS-12708 TS-12713 TS-12716 TS-12718 TS-12720 TS-12722
HYPETS-12712EA 0 EUR
HYPETS-12712 HYPETS-12708 HYPETS-12713 HYPETS-12716 HYPETS-12718 HYPETS-12720 HYPETS-12722
PTFE/silicone discs, Tuf-Bond™
Tussenschoten
These discs ensure air-tight, inert seals. They are specifically designed to combine the resealability of silicone with the inertness of PTFE.
  • Easily penetrated with standard syringe and CC needles
  • PTFE layer is 0,25 mm thick to ensure protection
These discs reseal instantly puncture after puncture, and the PTFE coating is structurally bonded so that no cement is leached or baked out of your sample after needle penetration.