Microcentrifuge tubes
Leverancier: BIOZYM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
710075 710080 710081 710090 710100 710101 710102 710103 710104 710105 710106 710108 710120 710121 710122 710123 710124 710125 710126 710128 710129 710135 710136 710157 710160 710161 710162 710163 710164 710165 710166 710168 710169 710175 710176 710180 710190 710191 710192 710193 710194 710195 710196 710198 710280 710300 710308 710310 710318 710320 710328 710340
BZYM710075EA 0 EUR
BZYM710075 BZYM710080 BZYM710081 BZYM710090 BZYM710100 BZYM710101 BZYM710102 BZYM710103 BZYM710104 BZYM710105 BZYM710106 BZYM710108 BZYM710120 BZYM710121 BZYM710122 BZYM710123 BZYM710124 BZYM710125 BZYM710126 BZYM710128 BZYM710129 BZYM710135 BZYM710136 BZYM710157 BZYM710160 BZYM710161 BZYM710162 BZYM710163 BZYM710164 BZYM710165 BZYM710166 BZYM710168 BZYM710169 BZYM710175 BZYM710176 BZYM710180 BZYM710190 BZYM710191 BZYM710192 BZYM710193 BZYM710194 BZYM710195 BZYM710196 BZYM710198 BZYM710280 BZYM710300 BZYM710308 BZYM710310 BZYM710318 BZYM710320 BZYM710328 BZYM710340
Microcentrifuge tubes
Buisjes Microcentrifugebuisjes
PE, translucent.
  • Good chemical resistance
  • Good temperature resistance
  • DNase-, RNase free
With or without cap.