Electroporation cuvettes
Leverancier: Biozym
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BZYM748040EA 0 EUR
BZYM748040 BZYM748041 BZYM748020 BZYM748021 BZYM748010 BZYM748011
Electroporation cuvettes
Cuvetten Elektroporatiecuvetten
These gamma sterilised electroporation cuvettes are suitable for all current electroporators.
  • Fitted with tight but easy to open closures
  • Gap width deviation inferior than 0,026 mm
  • Particularly smooth structured cuvette and electrode surfaces