Solid phase extraction cartridges, HyperSep™ C8
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
60108-309 60108-391 60108-392 60108-393 60108-394 60108-427 60108-704 60108-706
HYPE60108-309EA 181 EUR
HYPE60108-309 HYPE60108-391 HYPE60108-392 HYPE60108-393 HYPE60108-394 HYPE60108-427 HYPE60108-704 HYPE60108-706
Solid phase extraction cartridges, HyperSep™ C8
Kolommen voor chromatografie
HyperSep™ C8 cartridges, less retentive alternative to C18 for polar and nonpolar compounds.
  • Well suited for method requiring less retention than C18
  • For moderately polar analytes
  • Potential for polar interactions greater than with C18
HyperSep-96 system incorporates traditional solid phase extraction sorbents into a 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without methodMore Product Information