Capillary columns, with SafeGuard, TraceGOLD™ TG-1MS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26099-1425 26099-2985
555-9328EA 869 EUR
555-9328 555-9369
Capillary columns, with SafeGuard, TraceGOLD™ TG-1MS
Kolommen voor chromatografie
Exceptionally low bleed for optimal signal-to-noise ratio, sensitivity and MS integrity.
  • Non-polar
  • Equivalent to USP G2