Capillary columns, TRACE™ TR-35MS
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
260C286P 260C142P 260C154P
555-9304EA 519 EUR
555-9304 555-9311 555-9309
Capillary columns, TRACE™ TR-35MS
Kolommen voor chromatografie
Mid-polarity columns excellent for many applications.
  • Mid-polarity phase, 35% phenyl polysilphenylene-siloxane
  • Exceptionally low surface activity
  • Low bleed even at elevated temperatures