Fused silica tubing, Upchurch Scientific®
Leverancier: UPCHURCH SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FS-110 FS-115 FS-120 FS-150 FS-175
UPCHFS-110EA 260 EUR
UPCHFS-110 UPCHFS-115 UPCHFS-120 UPCHFS-150 UPCHFS-175
Fused silica tubing, Upchurch Scientific®
Slangen Chromatography Tubing
Made from high quality, polyimide-clad fused silica.
This tubing is ideal for micro-scale analyses such as micro and nano- HPLC and capillary electrophoresis because of the tight tolerances of fused silica’s inner diameters.