Screw caps and septa for Reacti-Vial™ and screw cap septum vials
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TS-13215 TS-13216 TS-13218 TS-13219
HYPETS-13215EA 61.8 EUR
HYPETS-13215 HYPETS-13216 HYPETS-13218 HYPETS-13219
Screw caps and septa for Reacti-Vial™ and screw cap septum vials
Sluitingen Sluitingen voor chromatografievials
These open top screw caps provide an inert, air-tight seal and direct puncture access to samples with a syringe needle.
  • Fits into Reacti-Vial™ small reaction vials
  • Used with PTFE/rubber laminated discs