Screw caps with valves, Mininert
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TS-10130 TS-10135
HYPETS-10130EA 268 EUR
HYPETS-10130 HYPETS-10135
Screw caps with valves, Mininert
Sluitingen Sluitingen voor chromatografievials
Mininert valves are screw caps that have integrated resealable valves to allow repeated and unlimited syringe-needle access to samples eliminates the septum-boring that occurs with repeated puncture of traditional septa. Made of chemical-resistant PTFE coating, they provide an inert, leak-tight, high-pressure seal.