Intestinaal epitheel differentiatiemedia, Corning®
Leverancier: Corning®
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Corning®
734-1433EA 305.33 EUR
734-1433
Intestinaal epitheel differentiatiemedia, Corning®
Media voor celscheiding
Corning® BioCoat™ Intestinal Epithelium Differentiation Environment is an integrated system designed to create in vitro intestinal models.
Includes 500mL Seeding Basal Medium, 500mL Differentiation Medium, and 2x0.5mL MITO+ Serum Extender.
Leveringsinformatie: Pakket bevat basaal entmedium (500 ml), differentiatiemedium (500 ml) en MITO+ Serum Extender (2×0,5 ml).