HPLC columns, Hypersil™ BDS Phenyl
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28905-254630 28903-154630 28905-254030 28905-154630 28905-304030
555-5909EA 926 EUR
555-5909 555-5936 555-5910 555-5952 555-5908
HPLC columns, Hypersil™ BDS Phenyl
Kolommen voor chromatografie
Hypersil™ BDS phenyl columns offer alternative selectively to Hypersil™ BDS C18 and C8 columns and phenyl phases offer clear advantages over alkyl chains when the compounds to be resolved. Used to accomplish the difficult separation.