Trolley, trays
Leverancier: Agencinox
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
445 745 441 947S 741 741S 746S 946S 441S 446S
139-0101EA 0 EUR
139-0101 139-0102 139-0100 AGEN947S AGEN741 AGEN741S AGEN746S AGEN946S AGEN441S AGEN446S
Trolley, trays
Transportwagens
Trolleys with two handles, shaped trays made up of 18/10 stainless steel with bright polish finishing.
  • Heavy duty structure
  • Easy cleaning
  • Available in knockdown construction or argon welded models