Heating tapes, IT-SiS10
Leverancier: ISOPAD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
115096-000 162874-000
632-4801EA 211.8 EUR
632-4801 ISOP162874-000
Heating tapes, IT-SiS10
Verwarmers
Isopad IT-SiS10 is a preterminated, flexible, and waterproof silicone insulated heating tape. Four silicone insulated heating conductors are embedded in a protective overbraid made of cupro-nickel. The temperature depends on the power and application.
  • Maximum operating temperature: 200 °C
  • Width×Thickness: 11×5 mm (±10%)
  • Maximum power per meter 100 W/m
It is non hazardous and used in ordinary areas. It is used for temperature modification of pipes and structures in industries and laboratories.