HPLC columns, Acclaim™ PolarAdvantage II (PA2)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
063189 063191 063197 063199 068973 069692 069699 071985 077997 077998 077999 078001 063705 066276 066277 063187
555-7327EA 626 EUR
555-7327 555-7328 555-7329 555-7330 555-7126 555-7120 555-7133 555-7182 555-7548 555-7549 555-7550 555-7552 555-7303 555-7155 555-7156 555-7326
HPLC columns, Acclaim™ PolarAdvantage II (PA2)
Kolommen voor chromatografie
These columns are used for the separation of both basic and acidic analytes, and incorporate amide-embedded polar group and multi point attachment between the ligands and the silica surface.
  • Selectively complementary to the C18 column
  • Compatible with 100% aqueous mobile phase
  • Wide range of applications
  • Low bleed for MS compatibility