Unions
Leverancier: VICI JOUR RESEARCH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
JR-065 JR-1064 JR-1065 JR-1068 JR-068 JR-064 JR-060-5
JOURJR-065EA 37.1 EUR
JOURJR-065 554-1026 554-1027 JOURJR-1068 JOURJR-068 JOURJR-064 JOURJR-060-5
Unions
Fittingen en connectoren
Unions for easy connection of low pressure solvent lines.
  • Low dead volume
  • Inert and biocompatible