Capillary columns, GuardGOLD™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
26050-0225 26050-0232 26050-0253 26050-0525 26050-0532 26050-0553 26050-1025 26050-1032 26050-1053
555-8854EA 62.4 EUR
555-8854 555-8853 555-8852 555-8850 555-8849 555-8825 555-8792 555-8791 555-8790
Capillary columns, GuardGOLD™
Kolommen voor chromatografie
These guard columns extend analytical column lifetime by preventing degradation from harsh 'steam cleaning' water injections.
  • Highly deactivated to provide superior inertness, essential for analysis of active compounds
  • Protects against column contamination caused by non-volatile materials