HPLC columns, Acclaim™ Explosives E2
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
069703 070081 070082 071989 064309
555-7135EA 378 EUR
555-7135 555-7199 555-7200 555-7183 555-7149
HPLC columns, Acclaim™ Explosives E2
Kolommen voor chromatografie
Acclaim™ Explosives E2 columns are used to resolve the nitroaromatics and nitramines, the novel and unique chemistries of these columns provide superior resolution with complementary selectivities.
  • Simple isocratic elution conditions
  • Rugged column with good lot-to-lot reproducibility