CombiPAL and GC PAL liquid injection syringes
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5188-6596 G6500-80113 G6500-80114 G6500-80115 G6500-80116 G6500-80117 G6500-80118 G6500-80119 G6500-80120
HEWL5188-6596EA 0 EUR
HEWL5188-6596 HEWLG6500-80113 HEWLG6500-80114 HEWLG6500-80115 HEWLG6500-80116 HEWLG6500-80117 HEWLG6500-80118 HEWLG6500-80119 HEWLG6500-80120
CombiPAL and GC PAL liquid injection syringes
Spuiten Injectiespuiten voor chromatografie
Liquid injection syringe system features with ability to inject a wide range of sample volumes up to 500 µl for LVI applications.
  • High reproducibility and accuracy
  • Flexible and with stringent quality
It has wide range of C-type syringes from 1,2 µl through 500 µl to accommodate fast and large-volume injections.