Replacement plungers
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G4200-80101 G4200-80103 G4200-80105 G4200-80107 G4200-80108 G4200-80111 G4200-80112
HEWLG4200-80101EA 0 EUR
HEWLG4200-80101 HEWLG4200-80103 HEWLG4200-80105 HEWLG4200-80107 HEWLG4200-80108 HEWLG4200-80111 HEWLG4200-80112
Replacement plungers
Spuitplunjer
Replacement plungers designed for long-lasting applications that demand precision and productivity.
  • Strongly recommended for high-throughput applications
  • X-type syringes feature zero carryover