Cellulose chromatography papers
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
817003A 817004A
MANA817003AEA 259 EUR
MANA817003A MANA817004A
Cellulose chromatography papers
Filters Filterpapier Chromatografiepapier
Paper chromatography is the chromatographic technique separation due to partition of the analytes between special paper grades and the mobile phase, which penetrates the paper by capillary action ascending, descending and circular techniques are possible.