HTS and HTC PAL liquid injection syringes
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5188-6485 G4200-80117 G4200-80118 G4200-80119
HEWL5188-6485EA 549 EUR
HEWL5188-6485 HEWLG4200-80117 HEWLG4200-80118 HEWLG4200-80119
HTS and HTC PAL liquid injection syringes
Spuiten Injectiespuiten voor chromatografie
These liquid injection syringes are designed specifically for CTC autosamplers.
These X-type syringes feature zero carryover and a long-lasting plunger for applications that demand precision and productivity. They are strongly recommended for high-throughput applications.