Gas detection system, Aerotest® Alpha
Leverancier: Dräger
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6527150 6527149
DRAE6527150EA 2210 EUR
DRAE6527150 DRAE6527149
Gas detection system, Aerotest® Alpha
Gasdetectiesystemen
Aerotest Alpha allows the quality of breathing air supplied by a compressor or compressed gas cylinder to be determined.
  • Efficient method of detecting and measuring toxic gases
  • Quick and easy application and independent of power source
  • Quantitative measurement of a variety of potentially harmful substances
The measurements can include carbon monoxide, carbon dioxide, water vapor and oil in the compressed air flow.