Drum taps, stainless steel
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2795 2797
MSGA2795EA 225 EUR
MSGA2795 MSGA2797
Drum taps, stainless steel
Kranen en gootstenen Kranen
These drum taps are made of stainless steel.
  • Adjustable removal position
  • Self closing
  • Flame guard
  • PTFE seal
Bestelinformatie: MSGA2797 includes a trailer fitting for decanting vessels.