Heating tapes
Leverancier: WINKLER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WBS00207300550 WBS00202700270 WBS00203100230 WBS00204000200 WBS00204000410 WBS00204600350 WBS00202200160 WBS00206300640 WBS00209500470 WBS00209500950 WBS00211000820 WBS00213500670 WBS00213501340 WBS00215501160 WBS00216000780 WBS00206300320 WBS00200900050 WBS00202700130 WBS00200900100 WBS00201100080 WBS00201800100 WBS00201800200 WBS00216001570 WBS00218301370 WBS00202900100
WINKWBS00207300550EA 0 EUR
WINKWBS00207300550 WINKWBS00202700270 WINKWBS00203100230 WINKWBS00204000200 WINKWBS00204000410 WINKWBS00204600350 WINKWBS00202200160 WINKWBS00206300640 WINKWBS00209500470 WINKWBS00209500950 WINKWBS00211000820 WINKWBS00213500670 WINKWBS00213501340 WINKWBS00215501160 WINKWBS00216000780 WINKWBS00206300320 WINKWBS00200900050 WINKWBS00202700130 WINKWBS00200900100 WINKWBS00201100080 WINKWBS00201800100 WINKWBS00201800200 WINKWBS00216001570 WINKWBS00218301370 WINKWBS00202900100
Heating tapes
Verwarmers
These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.
  • Very high and constant heating power per meter
  • High operating temperature
  • Extremely flexible
  • Quick and easy installation