Laser protection spectacles
Leverancier: DIDALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
POD020500 POD020510
DIDAPOD020500EA 83.2 EUR
DIDAPOD020500 DIDAPOD020510
Laser protection spectacles
Brillen
Single lens safety glasses protect against red and blue laser.
  • For wavelengths 600/760 nm or 200/540 nm
  • Goggles are adjustable, one size fits all