Bariumperchloraat 0.005 mol/l 2-propanol/water 80:20, Titripur® Reag. USP Volumetric, Supelco®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
MQ100
1.09086.1000 1.09086.1003
1.09086.1000EA 93.7 EUR
1.09086.1000 1.09086.1003
Bariumperchloraat 0.005 mol/l 2-propanol/water 80:20, Titripur® Reag. USP Volumetric, Supelco®
Bariumperchloraat
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.
Formule: Ba(ClO₄)₂
MW: 336.23 g/mol
Smeltpunt: 505
Vlampunt: 21
MDL Nummer: MFCD00003440
CAS nummer: 13465-95-7
UN: 1993
ADR: 3 II
Merck Index: 12,01016