Samples bags, isothermal
Leverancier: DELTALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
900074 900073 900076 900078
DELT900074EA 207 EUR
DELT900074 DELT900073 DELT900076 DELT900078
Samples bags, isothermal
Zakken Monsterzakken
These iothermal bags are used for the transport of biological samples, manufactured from PVC, nylon, LDPE, and polyurethane. They have two functions, i.e., keeping the temperature of samples from 8 to 12 H while being transported and to protect the samples and their contents.
  • Manufactured from sturdy and shockproof materials
  • Light and convenient with padlock
  • Space-saving to reduce volume by 50%
  • Corners are rounded and reinforced by a double seam
  • Handles are sewn all around the bag for increased strength
  • Three different types of locks, i.e., by velcro, by zip fastener and by a coded padlock (40 L)
They have an inner pocket designed to hold the cold packs, a transparent outer pocket, at the front of the bag, for laboratory identification and a transparent pocket, over the top of the bag, to insert documents.
Leveringsinformatie: Included cold packs (consist of HDPE packs filled with a non toxic gel).
Opgelet: Place the cold packs in the freezer and to be kept at −20 or 24 °C for 24 H before use.