Capillary columns, TRACE™ TR-1701
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
260Q142P 260Q143P 260Q298P
555-9401EA 732 EUR
555-9401 555-9400 555-9396
Capillary columns, TRACE™ TR-1701
Kolommen voor chromatografie
  • Mid-polarity phase, 14% cyanopropylphenyl polysiloxane
  • Low bleed even at a high operating temperature
  • Suitable for a wide variety of applications