Inoculating loops, Rotilabo®
Leverancier: Roth Carl
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
KL97.1 KN02.1
ROTHKL97.1EA 72.4 EUR
ROTHKL97.1 ROTHKN02.1
Inoculating loops, Rotilabo®
Uitstrijkspatels Entogen en -naalden
Rotilabo® inoculating loops, made of platinum/iridium. Wire diameter 0,6 mm. Rotilabo® inoculating loop holder set made of stainless steel. Loops are fixed with union nut. For inoculating loops with wire diameter 0,6 to 1,0 mm. Set sorted by colour: blue, yellow, red, black. Length: 245 mm. Autoclavable at 121 °C.
Leveringsinformatie: Loops delivered without inoculating loop holder, please order separately.