VWR®, Cover Slips, Rond
Leverancier: VWR Collection
Ratings:
Avg. Ratings: 3.0 (2 Reviews)
Total Ratings: 2
Avg. Ratings: 3.0 out of 5
Was this review helpful? Yes No
Yes: 5 No: 4 
Academia / Research associate / UK
Yes, I recommend
I used these coverslips for iPS derived neurons, and the cells attached very well (I pre-treated the coverslips with poly ornithine and matrigel), I used the same protocol for other coverslips, but in this one, the cells attached better and also looks better, with more neurites. I recommend this product.
Was this review helpful? Yes No
Yes: 5 No: 3 
Museum / Scientific Associate / London, UK
Yes, I recommend
I have been using 6mm coverslips for more than 40 years (oerioginal "Chance"). Over the past few years I have noticed a severe deterioration in the quality of the coverslips, but as they appear to be the only ones currently available I have been forced to carry on using them. The coverslips vary in diameter from 5.7-6.1mm and the edges are often very ragged. In about half of the coverslips the edges are not cleanly cut but have a piece remaining from the cutting process which usually needs to be broken off with a fine pair of forceps. Sometimes this piece is quite large. Compare these with the coverslips of 20-40 years ago which were of uniform diameter and with clean cut edges. I am only recommending them because they are the only suitable 6mm coverslips available.
631-0152EA 122 EUR
631-0152 631-0153 631-0162 631-0169 631-0170 631-0171 631-0172 631-0174 631-0176 631-0177 631-0178 631-0148 631-0149 631-0150 631-0154 631-0155 631-0158 631-0161 631-0168
VWR®, Cover Slips, Rond
Dekglaasjes
Gemaakt van zuiver wit Hydrolytisch klasse 1 glas.
  • Geen bubbels of gebreken
  • Schoongemaakt en gepolijst
Verpakking: in kunststof verdeeldozen met scharnierend deksel.