2-(Trifluoromethoxy)benzoic acid hydrazide
Leverancier: Thermo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2-(Trifluoromethoxy)benzhydrazide
SPB03521.1 SPB03521.10 SPB03521.50
MAYBSPB03521.1EA 0 EUR
MAYBSPB03521.1 MAYBSPB03521.10 MAYBSPB03521.50
2-(Trifluoromethoxy)benzoic acid hydrazide
2-(Trifluoromethoxy)benzoic acid hydrazide
Formule: C₈H₇F₃N₂O₂
MDL Nummer: MFCD00204180
CAS nummer: 175277-19-7