2-Bromo-4'-(difluoromethoxy)acetophenone

Leverancier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 4-(Difluoromethoxy)phenacyl bromide

TL00461DA TL00461EA
MAYBTL00461DAEA 0 EUR
MAYBTL00461DA MAYBTL00461EA
2-Bromo-4'-(difluoromethoxy)acetophenone
2-Bromo-4'-(difluoromethoxy)acetophenone
Formule: C₉H₇BrF₂O₂
MDL Nummer: MFCD00052200
CAS nummer: 141134-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR