D(-)-Luciferin sodium salt ≥95%
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Sodium (S)-2-(6-hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylate, D-Luciferin, sodium, D-Luciferin sodium, 4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid sodium salt
14682-50 14682-100 14682-500 14682-1
CAYM14682-50EA 57.1 EUR
CAYM14682-50 CAYM14682-100 CAYM14682-500 CAYM14682-1
D(-)-Luciferin sodium salt ≥95%
D(-)-Luciferin sodium salt
Formule: C₁₁H₇N₂NaO₃S₂
MW: 302.30 g/mol
MDL Nummer: MFCD00044938
CAS nummer: 103404-75-7